Anti-MDM2 Rabbit Polyclonal Antibody

Supplier: Prosci

70-666
10080-108EA 600.4 USD
10080-108
Anti-MDM2 Rabbit Polyclonal Antibody
Antibodies
MDM2 Antibody can be used for ELISA starting at 1:1000, and Immunohistochemistry in paraffin embeded tissues starting at 5 ?g/mL.

PBS, 0.09% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: MDM2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR