Anti-APOA1 Rabbit Polyclonal Antibody

Supplier: ProSci
70-647
10075-120EA 694.79 USD
10075-120
Anti-APOA1 Rabbit Polyclonal Antibody
Antibodies
APOA1 Antibody can be used for Radial Immunodiffusionstarting at 1:200, rocket IEP, Immunohistochemistry in paraffin embeded tissues starting at 1:100, and Western Blotting.

0.05% sodium azide, 100U/ml Penicillin, 10 ug/ml Streptomycin, 25 ng/ml Amphotericin B.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: APOA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Hamster, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR