Anti-SRSF3 Rabbit Polyclonal Antibody

Supplier: ProSci
70-641
10075-108EA 809.28 USD
10075-108
Anti-SRSF3 Rabbit Polyclonal Antibody
Antibodies
SRSF3 / SFRS3 Antibody can be used for ELISA starting at 1:5000, Immunofluorescence starting at 1:100 - 1:500, Immunohistochemistry in paraffin embeded tissues starting at 5?g/mL, and Western Blotting starting at 1:500 - 1:1000.

PBS (without Mg2+, Ca2+), pH 7.4, 150 mM NaCl, 0.02% sodium azide, 50% glycerol

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: SRSF3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR