Anti-FBN2 Rabbit Polyclonal Antibody

Supplier: ProSci
70-621
10073-988EA 719.12 USD
10073-988
Anti-FBN2 Rabbit Polyclonal Antibody
Antibodies
FBN2 / Fibrillin 2 Antibody can be used for ELISA starting at 1:1000 - 1:2000, Immunocytochemistry starting at 1:50 - 1:100, Immunohistochemistry in paraffin embeded tissues starting at 1:100, Immunoprecipitation 1:100 - 1:200, and Western Blotting starting at 1:200 - 1:500.

PBS

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: FBN2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR