Anti-AFP Rabbit Polyclonal Antibody

Supplier: ProSci
70-611
10077-544EA 694.79 USD
10077-544
Anti-AFP Rabbit Polyclonal Antibody
Antibodies
AFP / Alpha-Fetoprotein Antibody can be used for ELISA, Immunohistochemistry in paraffin embeded tissues starting at 2.5 ?g/mL, and Western Blotting.

PBS, pH 7.4, 0.02% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: AFP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR