Anti-HSPA1A Rabbit Polyclonal Antibody

Supplier: ProSci
70-610
10077-542EA 694.79 USD
10077-542
Anti-HSPA1A Rabbit Polyclonal Antibody
Antibodies
Heat Shock Protein 70 Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 2 ?g/mL, and Western Blotting.

PBS, pH 7.4, 0.02% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: HSPA1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR