Anti-CD81 Rabbit Polyclonal Antibody

Supplier: Prosci

70-604
10077-530EA 600.4 USD
10077-530
Anti-CD81 Rabbit Polyclonal Antibody
Antibodies
CD81 Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 2.5 ?g/mL, and Western Blotting starting at 1:1000 - 1:4000.

PBS, pH 7.4, 0.02% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CD81
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR