Anti-MRE11A Rabbit Polyclonal Antibody

Supplier: ProSci
70-602
10077-526EA 694.79 USD
10077-526
Anti-MRE11A Rabbit Polyclonal Antibody
Antibodies
MRE11A / MRE11 Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 10 ?g/mL, and Western Blotting starting at 1 - 2 ?g/mL.

PBS, 0.02 % sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: MRE11A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR