Anti-LY86 Rabbit Polyclonal Antibody

Supplier: ProSci
70-601
10075-084EA 773.05 USD
10075-084
Anti-LY86 Rabbit Polyclonal Antibody
Antibodies
LY86 / MD-1 Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 10 ?g/mL, and Immunoprecipitationstarting at 2 ?g/mL,

PBS, 0.05% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: LY86
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR