Anti-NDUFA13 Rabbit Polyclonal Antibody

Supplier: Prosci

70-598
10080-070EA 600.4 USD
10080-070
Anti-NDUFA13 Rabbit Polyclonal Antibody
Antibodies
NDUFA13 / GRIM19 Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 5 ?g/mL, and Western Blotting starting at 1:100 - 1:1000.

PBS containing 0.05% BSA and 0.05% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: NDUFA13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR