Anti-NOD1 Rabbit Polyclonal Antibody

Supplier: ProSci
70-585
10077-500EA 694.79 USD
10077-500
Anti-NOD1 Rabbit Polyclonal Antibody
Antibodies
NOD1 Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 1:100, and Western Blotting starting at 1:1000 - 1:2500.

50% glycerol, 0.025% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: NOD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR