Anti-TRAF3 Rabbit Polyclonal Antibody

Supplier: ProSci
70-581
10077-492EA 705.02 USD
10077-492
Anti-TRAF3 Rabbit Polyclonal Antibody
Antibodies
TRAF3 Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 10 ?g/mL, and Western Blotting starting at 1 - 3 ?g/mL.

PBS containing 0.05% BSA and 0.05% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: TRAF3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR