Anti-ANXA5 Rabbit Polyclonal Antibody

Supplier: ProSci
70-553
10073-940EA 705.02 USD
10073-940
Anti-ANXA5 Rabbit Polyclonal Antibody
Antibodies
ANXA5 / Annexin V Antibody can be used for ELISA starting at 1:1000 - 1:10000, and Immunohistochemistry in paraffin embeded tissues starting at 1:200,

PBS, pH 7.4, 0.02% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ANXA5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR