Anti-CDH23 Rabbit Polyclonal Antibody

Supplier: ProSci
70-550
10073-934EA 719.12 USD
10073-934
Anti-CDH23 Rabbit Polyclonal Antibody
Antibodies
CDH23 Antibody can be used for ELISA starting at 1:10000, and Immunofluorescence starting at 1:100 - 1:500,

PBS (without Mg2+, Ca2+), pH 7.4, 150 mM NaCl, 0.02% sodium azide, 50% glycerol

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CDH23
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR