Anti-LOC443301 Rabbit Polyclonal Antibody

Supplier: ProSci
70-549
10073-932EA 771.96 USD
10073-932
Anti-LOC443301 Rabbit Polyclonal Antibody
Antibodies
14-3-3 Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 1:200, Immunoprecipitation and Western Blotting starting at 1:2000.

0.09% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: LOC443301
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Plant, Amphibian, Bovine, Chicken, Fungus, Human, Mouse, Reptile, Rat, Sheep
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR