Anti-PLK1 Rabbit Polyclonal Antibody

Supplier: ProSci
70-548
10073-930EA 694.79 USD
10073-930
Anti-PLK1 Rabbit Polyclonal Antibody
Antibodies
PLK1 Antibody can be used for Immunohistochemistry, Immunohistochemistry in paraffin embeded tissues starting at 5 ?g/mL, and Western Blotting.

PBS, pH 7.4, 0.02% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: PLK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR