Anti-S100 A9 Rabbit Polyclonal Antibody

Supplier: Prosci

70-532
10073-898EA 600.4 USD
10073-898
Anti-S100 A9 Rabbit Polyclonal Antibody
Antibodies
S100A9 Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 10 ?g/mL, and Western Blotting starting at 1:200 - 1:2000.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: S100A9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR