Anti-HES5 Rabbit Polyclonal Antibody

Supplier: ProSci
70-521
10077-468EA 694.57 USD
10077-468
Anti-HES5 Rabbit Polyclonal Antibody
Antibodies
HES5 Antibody can be used for ELISA starting at 1:10000, and Immunohistochemistry in paraffin embeded tissues starting at 2.5 ?g/mL,

Tris-glycine (without Mg2+, Ca2+), pH 7.4, 0.02% sodium azide, 0.5% BSA

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: HES5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR