Anti-CDH13 Rabbit Polyclonal Antibody

Supplier: Prosci

70-503
10080-040EA 600.4 USD
10080-040
Anti-CDH13 Rabbit Polyclonal Antibody
Antibodies
CDH13 / H Cadherin Antibody can be used for ELISA starting at 1:1000, Flow Cytometry starting at 1:10 - 1:50, Immunohistochemistry in paraffin embeded tissues starting at 10?g/mL, and Western Blotting starting at 1:50 - 1:100.

PBS, 0.09% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CDH13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR