Anti-HPN Rabbit Polyclonal Antibody

Supplier: ProSci
70-479
10080-004EA 719.12 USD
10080-004
Anti-HPN Rabbit Polyclonal Antibody
Antibodies
HPN / Hepsin Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 5 ?g/mL, and Western Blotting starting at 1 ?g/mL.

TBS containing 50% glycerol, 0.5 mg/ml BSA, and 0.02% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: HPN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bat, Gibbon, Bovine, Dog, Gorilla, Human, Monkey, Orangutan, Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR