Anti-IGFBP5 Rabbit Polyclonal Antibody

Supplier: Prosci

70-474
10079-994EA 600.4 USD
10079-994
Anti-IGFBP5 Rabbit Polyclonal Antibody
Antibodies
IGFBP5 Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 5 ?g/mL, and Western Blotting starting at 1:200.

TBS pH 7.4, containing 50% glycerol, 0.5 mg/ml BSA, and 0.02% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: IGFBP5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bat, Gibbon, Bovine, Dog, Gorilla, Goat, Hamster, Horse, Human, Monkey, Pig, Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR