Anti-JUNB Rabbit Polyclonal Antibody

Supplier: ProSci
70-435
10075-062EA 705.02 USD
10075-062
Anti-JUNB Rabbit Polyclonal Antibody
Antibodies
JUNB Antibody can be used for ELISA starting at 1:20000, Immunohistochemistry in paraffin embeded tissues starting at 10 ?g/mL, Immunoprecipitation and Western Blotting starting at 1:500 - 1:1000.

PBS (without Mg2+, Ca2+), pH 7.4, 150 mM NaCl, 0.02% sodium azide, 50% glycerol

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: JUNB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR