Anti-SOD1 Rabbit Polyclonal Antibody

70-389
10079-858EA 694.79 USD
10079-858
Anti-SOD1 Rabbit Polyclonal Antibody
Antibodies
SOD1 Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 1:400, and Western Blotting starting at 1:2000,

HEPES with 0.15M NaCl, 0.01% BSA, 0.03% sodium azide, and 50% glycerol

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: SOD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR