Anti-CTTN Rabbit Polyclonal Antibody

70-373
10075-042EA 694.79 USD
10075-042
Anti-CTTN Rabbit Polyclonal Antibody
Antibodies
CTTN / Cortactin Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 1:200, and Western Blotting starting at 1:2000.

HEPES with 0.15M NaCl, 0.01% BSA, 0.03% sodium azide, and 50% glycerol

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CTTN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR