Anti-DDX4 Rabbit Polyclonal Antibody

70-354
10077-436EA 694.79 USD
10077-436
Anti-DDX4 Rabbit Polyclonal Antibody
Antibodies
DDX4 Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 5 ?g/mL.

PBS, 0.05% BSA, 0.05% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: DDX4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Chimpanzee, Dog, Horse, Human, Monkey, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR