Anti-HMGB3 Rabbit Polyclonal Antibody

70-348
10077-424EA 694.57 USD
10077-424
Anti-HMGB3 Rabbit Polyclonal Antibody
Antibodies
HMGB3 Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 10 ?g/mL, and Western Blotting starting at 0.5 - 1 ?g/mL.

PBS, 0.05% BSA, 0.05% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: HMGB3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Chimpanzee, Chicken, Dog, Horse, Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR