Anti-HSPA1A Chicken Polyclonal Antibody

70-335
10075-016EA 694.79 USD
10075-016
Anti-HSPA1A Chicken Polyclonal Antibody
Antibodies
HSP70 antibody can be used for staining paraffin embeded tissues starting at 5 ?g/mL, and Western Blot starting at 1:1000.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: HSPA1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR