Anti-GABRA1 Mouse Monoclonal Antibody [clone: [S96-55]]

Supplier: Prosci

70-334
10075-014EA 600.4 USD
10075-014
Anti-GABRA1 Mouse Monoclonal Antibody [clone: [S96-55]]
Antibodies
GABRB1 antibody can be used for immunocytochemistry starting at 0.1 - 1 ?g/mL, staining paraffin embeded tissues starting at 10 ?g/mL, immunoprecipitation starting at 1 - 10 ?g/mL, and Western Blot starting at 1 - 10 ?g/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: GABRA1
Clonality: Monoclonal
Clone: [S96-55]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR