Anti-KCNJ12 Mouse Monoclonal Antibody [clone: [S124B-38]]

70-329
10075-004EA 694.79 USD
10075-004
Anti-KCNJ12 Mouse Monoclonal Antibody [clone: [S124B-38]]
Antibodies
KCNJ12 antibody can be used for immunocytochemistry starting at 0.1 - 1 ?g/mL, staining paraffin embeded tissues starting at 5 ?g/mL, and Western Blot starting at 1 - 10 ?g/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: KCNJ12
Clonality: Monoclonal
Clone: [S124B-38]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR