Anti-FKBP4 Mouse Monoclonal Antibody

Supplier: Prosci

70-319
10079-824EA 600.4 USD
10079-824
Anti-FKBP4 Mouse Monoclonal Antibody
Antibodies
FKBP4 antibody can be used for staining paraffin embeded tissues starting at 10 ?g/mL, and Western Blot starting at 1:2000.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: FKBP4
Clonality: Monoclonal
Clone: [Hi52C]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Dog, Hamster, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR