Anti-HSP90AA1 Mouse Monoclonal Antibody [clone: [Hyb-K41009]]

Supplier: Prosci

70-315
10079-816EA 700.45 USD
10079-816
Anti-HSP90AA1 Mouse Monoclonal Antibody [clone: [Hyb-K41009]]
Antibodies
HSP90 Alpha antibody can be used for staining paraffin embeded tissues starting at 10 ?g/mL, and Western Blot starting at 1 ?g/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: HSP90AA1
Clonality: Monoclonal
Clone: [Hyb-K41009]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR