Anti-HSP90AB1 Mouse Monoclonal Antibody [clone: [H9010]]

Supplier: Prosci

70-314
10079-814EA 600.4 USD
10079-814
Anti-HSP90AB1 Mouse Monoclonal Antibody [clone: [H9010]]
Antibodies
  • HSP90 antibody can be used for staining paraffin embeded tissues starting at 10 μg/mL, and Western Blot starting at 1 μg/mL.

Protein G Column

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: HSP90AB1
Clonality: Monoclonal
Clone: [H9010]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Chicken, Dog, Human, Rabbit, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR