Anti-HSPA1A / HSPA1B Mouse Monoclonal Antibody [clone: [N27F3-4]]

70-313
10079-812EA 694.79 USD
10079-812
Anti-HSPA1A / HSPA1B Mouse Monoclonal Antibody [clone: [N27F3-4]]
Antibodies
HSP70 antibody can be used for staining paraffin embeded tissues starting at 10 ?g/mL, and Western Blot starting at 1 ?g/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: HSPA1A / HSPA1B
Clonality: Monoclonal
Clone: [N27F3-4]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Beluga, Bovine, C. elegans, Chicken, Dog, Drosophila, Fish, Xenopus, Guinea pig, Hamster, Human, Monkey, Mouse, Plant, Pig, Rabbit, Rat, Sheep
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR