Anti-RETN Mouse Monoclonal Antibody

70-312
10079-810EA 719.12 USD
10079-810
Anti-RETN Mouse Monoclonal Antibody
Antibodies
RETN antibody can be used for staining paraffin embeded tissues starting at 10 ?g/mL, and Western Blot starting at 0.2 - 0.4 ?g/mL.

Store Lyophilized at room temperature up to 1 month; Reconstituted for up to two weeks at 2-8°C. Aliquot and freeze at -20°C for long term storage. Avoid freeze/thaw cycles.

Type: Primary
Antigen: RETN
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR