Anti-UBB Mouse Monoclonal Antibody

Supplier: Prosci

70-304
10073-882EA 700.45 USD
10073-882
Anti-UBB Mouse Monoclonal Antibody
Antibodies
Ubiquitin antibody can be used for flow cytometry starting at 10 ?g/mL, staining paraffin embeded tissues starting at 10 ?g/mL, and Western Blot starting at 1:1000.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: UBB
Clonality: Monoclonal
Clone: [P4G7-H11]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Hamster, Human, Mouse, Rabbit, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR