Anti-Troponin I Type 2 Mouse Monoclonal Antibody [clone: [11E6]]

70-290
10079-798EA 705.02 USD
10079-798
Anti-Troponin I Type 2 Mouse Monoclonal Antibody [clone: [11E6]]
Antibodies
TNNI2 antibody can be used for staining paraffin embeded tissues starting at 10 ?g/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: TNNI2
Clonality: Monoclonal
Clone: [11E6]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR