Anti-TUBB2B Mouse Monoclonal Antibody

Supplier: Prosci

70-288
10073-860EA 578.75 USD
10073-860
Anti-TUBB2B Mouse Monoclonal Antibody
Antibodies
TUBB2B antibody can be used for staining paraffin embeded tissues starting at 5 ?g/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: TUBB2B
Clonality: Monoclonal
Clone: [AT5B3]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR