Anti-F8 Sheep Polyclonal Antibody

Supplier: Prosci
70-278
10073-842EA 809.28 USD
10073-842
Anti-F8 Sheep Polyclonal Antibody
Antibodies
F8 antibody can be used for staining paraffin embeded tissues starting at 5 ?g/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: F8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR