Anti-AHSA1 Rat Monoclonal Antibody [clone: [25F2.D10]]

Supplier: Prosci

70-277
10073-840EA 600.4 USD
10073-840
Anti-AHSA1 Rat Monoclonal Antibody [clone: [25F2.D10]]
Antibodies
AHSA1 antibody can be used for staining paraffin embeded tissues starting at 5 ?g/mL, and Western Blot starting at 1 ?g/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: AHSA1
Clonality: Monoclonal
Clone: [25F2.D10]
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a,k
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR