Anti-PPP2R5A Sheep Polyclonal Antibody

70-272
10073-832EA 694.79 USD
10073-832
Anti-PPP2R5A Sheep Polyclonal Antibody
Antibodies
Protein Phosphatase 2 A Antibody can be used for immunohistochemistry (tested on parafin embedded tissues) starting at 10 ?g/mL and Western Blot starting at 5 - 10 ?g/mL.

Provided as solution in phosphate buffered saline with 0.08% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: PPP2R5A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR