Anti-H3.1 Sheep Polyclonal Antibody

Supplier: Prosci

70-270
10073-828EA 600.4 USD
10073-828
Anti-H3.1 Sheep Polyclonal Antibody
Antibodies
Histone H3 Antibody can be used for immunocytochemistry starting at 1:100, immunohistochemistry (tested on parafin embedded tissues) starting at 5 ?g/mL and Western Blot starting at 1:300.

Phosphate buffered saline, pH 7.4

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: H3.1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Bovine, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR