Anti-F9 Sheep Polyclonal Antibody

Supplier: Prosci

70-269
10077-402EA 600.4 USD
10077-402
Anti-F9 Sheep Polyclonal Antibody
Antibodies
Factor 9 Antibody can be used for immunohistochemistry (tested on parafin embedded tissues) starting at 5 ?g/mL.

0.01 M HEPES, pH 7.4, 0.1 M NaCl, 50% glycerol.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: F9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR