Anti-SERPING1 Sheep Polyclonal Antibody

70-266
10077-398EA 718.9 USD
10077-398
Anti-SERPING1 Sheep Polyclonal Antibody
Antibodies
SERPING1 Antibody can be used for immunohistochemistry (tested on parafin embedded tissues) starting at 10 ?g/mL.

PBS, 0.09% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: SERPING1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Cat, Dog, Human, Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR