Anti-GFAP Chicken Polyclonal Antibody

70-265
10077-396EA 719.12 USD
10077-396
Anti-GFAP Chicken Polyclonal Antibody
Antibodies
GFAP Antibody can be used for immunocytochemistry starting at 1:100, immunofluorescence starting at 1:100 - 1:200, immunohistochemistry (tested on parafin embedded tissues) starting at 1:100 and Western Blot starting at 1:500.

PBS, pH 7.2, 0.02% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: GFAP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR