Anti-ACTA2 Mouse Monoclonal Antibody

70-257
10073-812EA 694.79 USD
10073-812
Anti-ACTA2 Mouse Monoclonal Antibody
Antibodies
Smooth Muscle Actin Monoclonal Antibody can be used for immunohistochemistry (tested on frozen tissues) starting at 1:10 - 1:50 and immunohistochemistry (tested on parafin embedded tissues) starting at 1:50.

PBS, pH 7.4, protein base, sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ACTA2
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR