Anti-Troponin C type 1 Mouse Monoclonal Antibody

70-254
10073-806EA 557.2 USD
10073-806
Anti-Troponin C type 1 Mouse Monoclonal Antibody
TNNC1 Monoclonal Antibody can be used for immunohistochemistry (tested on parafin embedded tissues) starting at 10 ?g/mL.

PBS, pH 7.2, 0.09% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: Troponin I
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Bovine, Cat, Dog, Fish, Goat, Human, Mouse, Pig, Rabbit, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR