Anti-C1QBP Mouse Monoclonal Antibody

70-253
10073-804EA 704.8 USD
10073-804
Anti-C1QBP Mouse Monoclonal Antibody
Antibodies
C1QBP Monoclonal Antibody can be used for immunohistochemistry (tested on parafin embedded tissues) starting at 1:50 and Western Blot starting at 1:2000 - 1:10000.

30% glycerol.

C1QBP Monoclonal antibody may be stored at 4°C; Avoid freeze-thaw cycles.

Type: Primary
Antigen: C1QBP
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR