Anti-S100 A8 Mouse Monoclonal Antibody [clone: [MAC387]]

Supplier: Prosci

70-252
10073-802EA 600.4 USD
10073-802
Anti-S100 A8 Mouse Monoclonal Antibody [clone: [MAC387]]
Antibodies
Macrophage Monoclonal Antibody can be used for flow cytometry starting at 1:50 - 1:100, immunohistochemistry (tested on frozen tissues starting at 1:100 - 1:200 and immunohistochemistry (tested on parafin embedded tissues) starting at 10 ?g/mL.

PBS containing 0.09% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: S100A8
Clonality: Monoclonal
Clone: [MAC387]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Cynomolgus monkey, Rhesus monkey, Baboon, Bovine, Cat, Dog, Guinea pig, Goat, Horse, Human, Pig, Rabbit, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR