Anti-PGRMC2 Mouse Monoclonal Antibody [clone: [3C11]]

70-235
10079-792EA 622.74 USD
10079-792
Anti-PGRMC2 Mouse Monoclonal Antibody [clone: [3C11]]
PGRMC2 Monoclonal Antibody can be used for immunohistochemistry (tested on parafin embedded tissues) starting at 1 - 10 ?g/mL and Western Blot starting at 1:500 - 1:1000.

PBS, pH 7.2

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: PGRMC2
Clonality: Monoclonal
Clone: [3C11]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1,k
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR