Anti-F9 Mouse Monoclonal Antibody [clone: [13F42-F6]]

70-228
10075-002EA 719.12 USD
10075-002
Anti-F9 Mouse Monoclonal Antibody [clone: [13F42-F6]]
Antibodies
F9 Monoclonal Antibody can be used for immunohistochemistry (tested on parafin embedded tissues) starting at 10 ?g/mL.

0.05M Sodium Phosphate; 0.1M NaCl; 1mM EDTA; pH 6.6

F9 Monoclonal antibody may be stored at Store at -70°C

Type: Primary
Antigen: F9
Clonality: Monoclonal
Clone: [13F42-F6]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR